3-Bromo-4-chloro-5-methylpyridine 95%

Lieferant: Apollo Scientific
Achtung

OR500016-1G OR500016-250MG
APOSOR500016-1GEA 451 EUR
APOSOR500016-1G APOSOR500016-250MG
3-Bromo-4-chloro-5-methylpyridine 95%
3-Bromo-4-chloro-5-methylpyridine
Formel: C₆H₅BrClN
Molecular Weight: 206.468 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD18415475
CAS-Nummer: 1261786-46-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR