2-Bromo-4-methylnicotinonitrile 95+%

Lieferant: Apollo Scientific
Achtung

OR500018-250MG
APOSOR500018-250MGEA 94.4 EUR
APOSOR500018-250MG
2-Bromo-4-methylnicotinonitrile 95+%
2-Bromo-4-methylnicotinonitrile
Formel: C₇H₅BrN₂
Molecular Weight: 197.032 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09834634
CAS-Nummer: 65996-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR