5-Cyclopropylpicolinic acid hydrochloride 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR500019-100MG
OR500019-250MG
APOSOR500019-100MGEA
95.7
EUR
APOSOR500019-100MG
APOSOR500019-250MG
5-Cyclopropylpicolinic acid hydrochloride 97%
5-Cyclopropylpicolinic acid hydrochloride
Formel:
C₉H₉NO₂·ClH Lagertemperatur: Raumtemperatur |
MDL:
MFCD30730075 CAS-Nummer: 2089378-52-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...