7-Bromo-2-chloroquinazolin-4-amine 95+%

Lieferant: Apollo Scientific
Achtung

OR500022-250MG
APOSOR500022-250MGEA 56.7 EUR
APOSOR500022-250MG
7-Bromo-2-chloroquinazolin-4-amine 95+%
7-Bromo-2-chloroquinazolin-4-amine
Formel: C₈H₅BrClN₃
Molecular Weight: 258,502 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11858276
CAS-Nummer: 1107695-08-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR