7-Chloroquinolin-8-ol 97%

Lieferant: Apollo Scientific
Achtung

OR500023-250MG
APOSOR500023-250MGEA 120 EUR
APOSOR500023-250MG
7-Chloroquinolin-8-ol 97%
7-Chloroquinolin-8-ol
Formel: C₉H₆ClNO
Molecular Weight: 179,603 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11870758
CAS-Nummer: 876-86-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR