5-Bromo-1H-imidazole-4-carbaldehyde 95+%

Lieferant: Apollo Scientific
Achtung

OR500024-250MG
APOSOR500024-250MGEA 125 EUR
APOSOR500024-250MG
5-Bromo-1H-imidazole-4-carbaldehyde 95+%
5-Bromo-1H-imidazole-4-carbaldehyde
Formel: C₄H₃BrN₂O
Molecular Weight: 174,983 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD21336623
CAS-Nummer: 50743-01-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR