2-Chloroquinoline-6-carbaldehyde 97%

Lieferant: Apollo Scientific
Achtung

OR500027-100MG OR500027-250MG
APOSOR500027-100MGEA 109 EUR
APOSOR500027-100MG APOSOR500027-250MG
2-Chloroquinoline-6-carbaldehyde 97%
2-Chloroquinoline-6-carbaldehyde
Formel: C₁₀H₆ClNO
Molecular Weight: 191.614 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08236769
CAS-Nummer: 791626-59-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR