2-Chloroquinoline-6-carbaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR500027-100MG
OR500027-250MG
APOSOR500027-100MGEA
109
EUR
APOSOR500027-100MG
APOSOR500027-250MG
2-Chloroquinoline-6-carbaldehyde 97%
2-Chloroquinoline-6-carbaldehyde
Formel:
C₁₀H₆ClNO Molecular Weight: 191.614 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD08236769 CAS-Nummer: 791626-59-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...