7-Bromobenzo[d]thiazol-2(3H)-one 95+%

Lieferant: Apollo Scientific
Achtung

OR500033-250MG
APOSOR500033-250MGEA 130 EUR
APOSOR500033-250MG
7-Bromobenzo[d]thiazol-2(3H)-one 95+%
7-Bromobenzo[d]thiazol-2(3H)-one
Formel: C₇H₄BrNOS
Molecular Weight: 230.082 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09749232
CAS-Nummer: 1188047-07-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR