7-Bromoimidazo[1,2-a]pyrimidine 97%

Lieferant: Apollo Scientific
Achtung

OR500037-250MG
APOSOR500037-250MGEA 444 EUR
APOSOR500037-250MG
7-Bromoimidazo[1,2-a]pyrimidine 97%
7-Bromoimidazo[1,2-a]pyrimidine
Formel: C₆H₄BrN₃
Lagertemperatur: Gefrierschrank
MDL: MFCD16877232
CAS-Nummer: 1251033-57-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR