3-tert-Butyl-1-methyl-2-pyrazolin-5-one

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-(tert-Butyl)-2,4-dihydro-2-methyl-3H-pyrazol-3-one

OR5005-1G
APOSOR5005-1GEA 87 EUR
APOSOR5005-1G
3-tert-Butyl-1-methyl-2-pyrazolin-5-one
3-tert-Butyl-1-methyl-2-pyrazolin-5-one
Formel: C₈H₁₄N₂O
Molecular Weight: 154,21 g/mol
Schmelzpunkt: 147…151 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051761
CAS-Nummer: 87031-30-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR