2,6-Dimethoxybenzonitril 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR5007-10G
APOSOR5007-10GEA
51
EUR
APOSOR5007-10G
2,6-Dimethoxybenzonitril 97%
2,6-Dimethoxybenzonitril
Formel:
C₉H₉NO₂ Schmelzpunkt: 114…116 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001788 CAS-Nummer: 16932-49-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...