2-Acetoxybenzonitril 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Cyanophenyl acetate

OR5008-25G
APOSOR5008-25GEA 132 EUR
APOSOR5008-25G
2-Acetoxybenzonitril 98%
2-Acetoxybenzonitril
Formel: C₉H₇NO₂
Molecular Weight: 161,16 g/mol
Siedepunkt: 119…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017360
CAS-Nummer: 5715-02-6
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR