4-Chlormethyl-o-xylol 70%
Lieferant: Apollo Scientific
OR5034-100G
OR5034-10G
OR5034-25G
APOSOR5034-100GEA
237
EUR
APOSOR5034-100G
APOSOR5034-10G
APOSOR5034-25G
4-Chlormethyl-o-xylol 70%
4-Chlormethyl-o-xylol
Formel:
C₉H₁₁Cl Siedepunkt: 117 °C/24mmHg Dichte: 1,06 g/cm³ (25 °C) |
MDL:
MFCD00000910 CAS-Nummer: 102-46-5 EINECS: 203-032-5 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...