Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%

Lieferant: Apollo Scientific

Synonyme: (R)-1-tert-Butyl 2-methyl 4-oxopiperidine-1,2-dicarboxylate

OR50476-1G
APOSOR50476-1GEA 232 EUR
APOSOR50476-1G
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected 95%
Methyl (2R)-4-oxopiperidine-2-carboxylate, N-BOC protected
Formel: C₁₂H₁₉NO₅
Molecular Weight: 257,283 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD12547038
CAS-Nummer: 1799811-83-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR