[(2S)-5-Oxo-2-pyrrolidinyl]acetic acid 95%

Lieferant: Apollo Scientific

OR50627-1G
APOSOR50627-1GEA 536 EUR
APOSOR50627-1G
[(2S)-5-Oxo-2-pyrrolidinyl]acetic acid 95%
[(2S)-5-Oxo-2-pyrrolidinyl]acetic acid
Formel: C₆H₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD19690753
CAS-Nummer: 61884-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR