2-Ethoxynicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

OR5067-5G
APOSOR5067-5GEA 68.3 EUR
APOSOR5067-5G
2-Ethoxynicotinsäure 98%
2-Ethoxynicotinsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Schmelzpunkt: 90…92 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00128788
CAS-Nummer: 35969-54-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR