2-Hydroxynicotinsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxypyridin-3-carbonsäure, 1,2-Dihydro-2-oxonicotinsäure, 2-Hydroxy-3-pyridincarbonsäure

OR5073-1G
APOSOR5073-1GEA 14.7 EUR
APOSOR5073-1G
2-Hydroxynicotinsäure 98%
2-Hydroxynicotinsäure
Formel: C₆H₅NO₃
Molecular Weight: 139,11 g/mol
Schmelzpunkt: 258…261 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010100
CAS-Nummer: 609-71-2
EINECS: 210-198-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR