3-Brom-5-methylanisol 97%

Lieferant: Apollo Scientific

Synonyme: 3-Bromo-5-methoxytoluene, 1-Bromo-3-methoxy-5-methylbenzene

OR50833-25G
APOSOR50833-25GEA 63.8 EUR
APOSOR50833-25G
3-Brom-5-methylanisol 97%
3-Brom-5-methylanisol
Formel: C₈H₉BrO
Molecular Weight: 201,0600 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08061916
CAS-Nummer: 29578-83-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR