1,3-Oxazolo[4,5-b]pyridin-2(3H)thione 98%

Lieferant: Apollo Scientific

Synonyme: 2,3-Dihydro-2-thioxo-1,3-oxazolo[4,5-b]pyridine, Oxazolo[4,5-b]pyridine-2(3H)-thione

OR5092-5G
APOSOR5092-5GEA 87 EUR
APOSOR5092-5G
1,3-Oxazolo[4,5-b]pyridin-2(3H)thione 98%
1,3-Oxazolo[4,5-b]pyridin-2(3H)thione
Formel: C₆H₄N₂OS
Schmelzpunkt: 243…246 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01089040
CAS-Nummer: 53052-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR