2-Ethoxyacetyl chloride 97%

Lieferant: Apollo Scientific

OR50944-100G OR50944-25G OR50944-5G
APOSOR50944-100GEA 0 EUR
APOSOR50944-100G APOSOR50944-25G APOSOR50944-5G
2-Ethoxyacetyl chloride 97%
2-Ethoxyacetyl chloride
Formel: C₄H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11170566
CAS-Nummer: 14077-58-8
UN: 2920
ADR: 8+3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR