5,5'-Dibrom-2,2'-bipyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5,5'-Dibromo-2,2'-bipyridine

OR50951-1G OR50951-5G
APOSOR50951-1GEA 125 EUR
APOSOR50951-1G APOSOR50951-5G
5,5'-Dibrom-2,2'-bipyridin
5,5'-Dibrom-2,2'-bipyridin
Formel: C₁₀H₆Br₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD06657883
CAS-Nummer: 15862-18-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR