3,4-(Trimethylenedioxy)benzoic acid 95%
Lieferant: Apollo Scientific
Synonyme:
3,4-Dihydro-2H-benzo[b][1,4]dioxepine-2-carboxylic acid
OR50957-1G
APOSOR50957-1GEA
121
EUR
APOSOR50957-1G
3,4-(Trimethylenedioxy)benzoic acid 95%
3,4-(Trimethylenedioxy)benzoic acid
Formel:
C₁₀H₁₀O₄ Molecular Weight: 194.18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09953183 CAS-Nummer: 33632-74-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...