4-(2,2-Dimethylpropanoylamino)benzenesulphonyl chloride 95%

Lieferant: Apollo Scientific

OR50987-1G OR50987-250MG
APOSOR50987-1GEA 647 EUR
APOSOR50987-1G APOSOR50987-250MG
4-(2,2-Dimethylpropanoylamino)benzenesulphonyl chloride 95%
4-(2,2-Dimethylpropanoylamino)benzenesulphonyl chloride
Formel: C₁₁H₁₄ClNO₃S
Molecular Weight: 275.752 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09971327
CAS-Nummer: 438191-85-6
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR