3-Bromoprop-1-en-2-ylbenzene

Lieferant: Apollo Scientific

OR50994-100MG
APOSOR50994-100MGEA 128 EUR
APOSOR50994-100MG
3-Bromoprop-1-en-2-ylbenzene
3-Bromoprop-1-en-2-ylbenzene
Formel: C₉H₉Br
Molecular Weight: 197,072 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09031921
CAS-Nummer: 3360-54-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR