(R)-(4-Methylmorpholin-3-yl)methanol ≥97%

Lieferant: Apollo Scientific

OR510000-250MG
APOSOR510000-250MGEA 318 EUR
APOSOR510000-250MG
(R)-(4-Methylmorpholin-3-yl)methanol ≥97%
(R)-(4-Methylmorpholin-3-yl)methanol
Formel: C₆H₁₃NO₂
Lagertemperatur: Kühlschrank
MDL: MFCD26792468
CAS-Nummer: 1620510-51-1
UN: 1760
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR