2-Chloro-6-isopropylpyridine > 97%
Lieferant: Apollo Scientific
OR510004-250MG
APOSOR510004-250MGEA
71.2
EUR
APOSOR510004-250MG
2-Chloro-6-isopropylpyridine > 97%
2-Chloro-6-isopropylpyridine
Formel:
C₈H₁₀ClN Molecular Weight: 155.625 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD08689614 CAS-Nummer: 120145-22-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...