2-Methyl-1h-indole-5-carbonitrile ≥95%

Lieferant: Apollo Scientific

OR510005-250MG
APOSOR510005-250MGEA 224 EUR
APOSOR510005-250MG
2-Methyl-1h-indole-5-carbonitrile ≥95%
2-Methyl-1h-indole-5-carbonitrile
Formel: C₁₀H₈N₂
Molecular Weight: 156,184 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09263860
CAS-Nummer: 36798-24-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR