3-Methylquinolin-8-amine ≥96%
Lieferant: Apollo Scientific
OR510007-250MG
APOSOR510007-250MGEA
331
EUR
APOSOR510007-250MG
3-Methylquinolin-8-amine ≥96%
3-Methylquinolin-8-amine
Formel:
C₁₀H₁₀N₂ Molecular Weight: 158,2 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD18449011 CAS-Nummer: 3393-71-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...