(6-Methylpyridin-2-yl)methanamine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510008-1G
APOSOR510008-1GEA 48 EUR
APOSOR510008-1G
(6-Methylpyridin-2-yl)methanamine hydrochloride ≥98%
(6-Methylpyridin-2-yl)methanamine hydrochloride
Formel: C₇H₁₀N₂·ClH
Molecular Weight: 158,629 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09878926
CAS-Nummer: 1365836-53-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR