6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine ≥97%

Lieferant: Apollo Scientific

OR510011-250MG
APOSOR510011-250MGEA 84.1 EUR
APOSOR510011-250MG
6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine ≥97%
6-Chloro-1-methyl-1H-pyrazolo[4,3-c]pyridine
Formel: C₇H₆ClN₃
Molecular Weight: 167,596 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD28608322
CAS-Nummer: 1558302-68-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR