4-Bromo-1H-pyrrole-2-carbonitrile ≥95%
Lieferant: Apollo Scientific
OR510013-250MG
APOSOR510013-250MGEA
58
EUR
APOSOR510013-250MG
4-Bromo-1H-pyrrole-2-carbonitrile ≥95%
4-Bromo-1H-pyrrole-2-carbonitrile
Formel:
C₅H₃BrN₂ Molecular Weight: 170,995 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD20502410 CAS-Nummer: 1221435-18-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...