1-(2-Chloroethyl)piperidine-4-carbonitrile ≥97%
Lieferant: Apollo Scientific
OR510014-250MG
APOSOR510014-250MGEA
336
EUR
APOSOR510014-250MG
1-(2-Chloroethyl)piperidine-4-carbonitrile ≥97%
1-(2-Chloroethyl)piperidine-4-carbonitrile
Formel:
C₈H₁₃ClN₂ Molecular Weight: 172,655 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD04108054 CAS-Nummer: 108890-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...