3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥98%

Lieferant: Apollo Scientific

OR510015-250MG
APOSOR510015-250MGEA 125 EUR
APOSOR510015-250MG
3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride ≥98%
3-Methyl-5,6,7,8-tetrahydroimidazo[1,5-a]pyrazine hydrochloride
Formel: C₇H₁₁N₃·ClH
Molecular Weight: 173,643 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15072031
CAS-Nummer: 601515-50-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR