2-Chloroquinolin-7-ol ≥95%
Lieferant: Apollo Scientific
OR510016-250MG
APOSOR510016-250MGEA
58
EUR
APOSOR510016-250MG
2-Chloroquinolin-7-ol ≥95%
2-Chloroquinolin-7-ol
Formel:
C₉H₆ClNO Molecular Weight: 179,603 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD02238180 CAS-Nummer: 375358-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...