5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde ≥97%

Lieferant: Apollo Scientific

OR510017-250MG
APOSOR510017-250MGEA 106 EUR
APOSOR510017-250MG
5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde ≥97%
5-Chloroimidazo[1,2-a]pyridine-3-carbaldehyde
Formel: C₈H₅ClN₂O
Lagertemperatur: Kühlschrank
CAS-Nummer: 198895-50-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR