5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde ≥97%

Lieferant: Apollo Scientific

OR510018-250MG
APOSOR510018-250MGEA 53.8 EUR
APOSOR510018-250MG
5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde ≥97%
5-Chloroimidazo[1,2-a]pyridine-2-carbaldehyde
Formel: C₈H₅ClN₂O
Lagertemperatur: Kühlschrank
CAS-Nummer: 881841-35-6
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR