4-Chloro-2H-chromen-2-one ≥98%

Lieferant: Apollo Scientific

OR510019-250MG
APOSOR510019-250MGEA 66.7 EUR
APOSOR510019-250MG
4-Chloro-2H-chromen-2-one ≥98%
4-Chloro-2H-chromen-2-one
Formel: C₉H₅ClO₂
Molecular Weight: 180,588 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00463528
CAS-Nummer: 17831-88-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR