1,2,3,4-Tetrahydro-1,4-epiminonaphthalene hydrochloride ≥95%
Lieferant: Apollo Scientific
OR510020-250MG
APOSOR510020-250MGEA
241
EUR
APOSOR510020-250MG
1,2,3,4-Tetrahydro-1,4-epiminonaphthalene hydrochloride ≥95%
1,2,3,4-Tetrahydro-1,4-epiminonaphthalene hydrochloride
Formel:
C₁₀H₁₁N·ClH Molecular Weight: 181,662 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD28125210 CAS-Nummer: 5176-31-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...