4-Aminopyridin-3-ol dihydrochloride ≥95%

Lieferant: Apollo Scientific

OR510021-1G
APOSOR510021-1GEA 50.9 EUR
APOSOR510021-1G
4-Aminopyridin-3-ol dihydrochloride ≥95%
4-Aminopyridin-3-ol dihydrochloride
Formel: C₅H₆N₂O·2ClH
Molecular Weight: 183,036 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 1081776-23-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR