5-Formyl-4-methyl-1H-indole-2-carbonitrile ≥97%

Lieferant: Apollo Scientific

OR510022-250MG
APOSOR510022-250MGEA 134 EUR
APOSOR510022-250MG
5-Formyl-4-methyl-1H-indole-2-carbonitrile ≥97%
5-Formyl-4-methyl-1H-indole-2-carbonitrile
Formel: C₁₁H₈N₂O
Molecular Weight: 184,194 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD31567509
CAS-Nummer: 1857296-22-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR