3-Bromo-4-hydrazinylpyridine > 97%

Lieferant: Apollo Scientific

OR510023-250MG
APOSOR510023-250MGEA 87 EUR
APOSOR510023-250MG
3-Bromo-4-hydrazinylpyridine > 97%
3-Bromo-4-hydrazinylpyridine
Formel: C₅H₆BrN₃
Molecular Weight: 188.025 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00234090
CAS-Nummer: 1202941-04-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR