4-Bromo-2-isopropylpyridine ≥97%

Lieferant: Apollo Scientific

OR510026-250MG
APOSOR510026-250MGEA 323 EUR
APOSOR510026-250MG
4-Bromo-2-isopropylpyridine ≥97%
4-Bromo-2-isopropylpyridine
Formel: C₈H₁₀BrN
Molecular Weight: 200.076 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD11223217
CAS-Nummer: 908267-63-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR