5-Bromo-2-methylbenzyl alcohol ≥97%

Lieferant: Apollo Scientific

OR510027-250MG
APOSOR510027-250MGEA 0 EUR
APOSOR510027-250MG
5-Bromo-2-methylbenzyl alcohol ≥97%
5-Bromo-2-methylbenzyl alcohol
Formel: C₈H₉BrO
Lagertemperatur: Kühlschrank
MDL: MFCD12031921
CAS-Nummer: 258886-04-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR