2-(Isopropylthio)aniline hydrochloride ≥98%
Lieferant: Apollo Scientific
OR510028-250MG
APOSOR510028-250MGEA
0
EUR
APOSOR510028-250MG
2-(Isopropylthio)aniline hydrochloride ≥98%
2-(Isopropylthio)aniline hydrochloride
Formel:
C₉H₁₃NS·ClH Lagertemperatur: Kühlschrank |
MDL:
MFCD11841393 CAS-Nummer: 861343-73-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...