2-Chloroquinoline-8-carboxylic acid ≥95%
Lieferant: Apollo Scientific
OR510029-250MG
APOSOR510029-250MGEA
323
EUR
APOSOR510029-250MG
2-Chloroquinoline-8-carboxylic acid ≥95%
2-Chloroquinoline-8-carboxylic acid
Formel:
C₁₀H₆ClNO₂ Molecular Weight: 207,613 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD11557275 CAS-Nummer: 1092287-54-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...