4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride ≥96%

Lieferant: Apollo Scientific

OR510030-250MG
APOSOR510030-250MGEA 143 EUR
APOSOR510030-250MG
4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride ≥96%
4,5,6,7-Tetrahydro-1H-indazol-5-amine dihydrochloride
Formel: C₇H₁₁N₃·2ClH
Molecular Weight: 210,104 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09028167
CAS-Nummer: 74197-17-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR