5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine ≥98%

Lieferant: Apollo Scientific

OR510032-250MG
APOSOR510032-250MGEA 79.9 EUR
APOSOR510032-250MG
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine ≥98%
5-Bromo-1-methyl-1H-pyrazolo[3,4-c]pyridine
Formel: C₇H₆BrN₃
Molecular Weight: 212,047 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD22380195
CAS-Nummer: 1337880-46-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR