7-Bromo-2,3-dihydro-1H-inden-1-ol ≥97%

Lieferant: Apollo Scientific

OR510033-250MG
APOSOR510033-250MGEA 188 EUR
APOSOR510033-250MG
7-Bromo-2,3-dihydro-1H-inden-1-ol ≥97%
7-Bromo-2,3-dihydro-1H-inden-1-ol
Formel: C₉H₉BrO
Molecular Weight: 213,071 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD24843663
CAS-Nummer: 1196049-18-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR