2-Bromo-3-methylisonicotinic acid ≥97%

Lieferant: Apollo Scientific

OR510034-250MG
APOSOR510034-250MGEA 139 EUR
APOSOR510034-250MG
2-Bromo-3-methylisonicotinic acid ≥97%
2-Bromo-3-methylisonicotinic acid
Formel: C₇H₆BrNO₂
Molecular Weight: 216,032 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD16610192
CAS-Nummer: 1211583-05-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR